Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α-Trifluoro-o-toluonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-(Trifluoromethyl)benzonitrileo-(Trifluoromethyl)benzonitrile2-Cyanobenzotrifluoride

PC7550-25G
77418-002EA 21 USD InStock
77418-002
α,α,α-Trifluoro-o-toluonitrile 98%
α,α,α-Trifluoro-o-toluonitrile


Formula: C₈H₄F₃N
MW: 171.12 g/mol
Boiling Pt: 204…206 °C
Melting Pt: 17…19 °C
Density: 1.294 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00001791
CAS Number: 447-60-9
EINECS: 207-184-3
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty