Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Piperidin-1-yl)benzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR0759-100MG
77402-218EA 13 USD InStock
77402-218
4-(Piperidin-1-yl)benzoic acid
4-(Piperidin-1-yl)benzoic acid


Formula: C₁₂H₁₅NO₂
MW: 205.2530 g/mol
Melting Pt: 214…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00587602
CAS Number: 22090-24-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty