Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-4-nitroaniline 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR3796-500G OR3796-100G OR3796-25G
77409-370EA 13 USD InStock
77409-370 77409-372 77409-368
N-Methyl-4-nitroaniline 97%
N-Methyl-4-nitroaniline


Formula: C₇H₈N₂O₂
Melting Pt: 149…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00007305
CAS Number: 100-15-2
EINECS: 202-823-2
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty