Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fmoc-Hyp-OH 98%

Supplier: APOLLO SCIENTIFIC
Synonyms: N-Fmoc-trans-4-hydroxy-L-proline

OR307070-100G OR307070-25G
77419-032EA 181 USD InStock
77419-032 77419-034
Fmoc-Hyp-OH 98%
Fmoc-Hyp-OH


Formula: C₂₀H₁₉NO₅
MW: 353.374 g/mol
Melting Pt: 189…193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151929
CAS Number: 88050-17-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty