Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-1-(Boc-amino)-2-propanol 98%

Supplier: APOLLO SCIENTIFIC

OR307132-100G OR307132-25G OR307132-5G
77419-044EA 318 USD InStock
77419-044 77419-048 77419-046
(S)-1-(Boc-amino)-2-propanol 98%
(S)-1-(Boc-amino)-2-propanol


Formula: C₈H₁₇NO₃
MW: 175.23
Storage Temperature: Ambient
MDL Number: MFCD04974340
CAS Number: 167938-56-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty