Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-3-chloropyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR30948-25G OR30948-100G
77419-270EA 23 USD InStock
77419-270 77419-268
2-Amino-3-chloropyridine 98%
2-Amino-3-chloropyridine


Formula: C₅H₅ClN₂
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD03541052
CAS Number: 39620-04-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty