Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Proline 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (±)-ProlineH-DL-Pro-OH

OR350110-500G OR350110-1KG OR350110-100G
77420-426EA 248 USD InStock
77420-426 77420-428 77420-424
DL-Proline 98%
DL-Proline


Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 208…210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005250
CAS Number: 609-36-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty