Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrochalcone

Supplier: APOLLO SCIENTIFIC
Warning

OR3872-25G OR3872-5G
77420-770EA 100 USD InStock
77420-770 77420-768
2-Nitrochalcone
2-Nitrochalcone


Formula: C₁₅H₁₁NO₃
MW: 253.26 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00031069
CAS Number: 7473-93-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty