Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Tolualdehyde 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: o-Tolualdehyde2-Methylbenzaldehyde

OR46026-500G OR46026-100G OR46026-25G
77421-758EA 13 USD InStock
77421-758 77421-760 77421-756
2-Tolualdehyde 98%
2-Tolualdehyde


Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 199…200 °C
Melting Pt: –35 °C
Density: 1.037 g/cm³ (25 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00003338
CAS Number: 529-20-4
EINECS: 208-452-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty