Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Denatonium benzoate 98%

Supplier: APOLLO SCIENTIFIC
Synonyms: N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate

OR50488-100G
77422-362EA 70 USD InStock
77422-362
Denatonium benzoate 98%
Denatonium benzoate


Formula: C₂₈H₃₄N₂O₃
MW: 446.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031578
CAS Number: 3734-33-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty