Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diglycolic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2,2'-Oxydiacetic acid

OR55403-100G OR55403-500G
77423-058EA 121 USD InStock
77423-058 77423-060
Diglycolic acid 98%
Diglycolic acid


Formula: C₄H₆O₅
MW: 134.09 g/mol
Melting Pt: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00004309
CAS Number: 110-99-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty