Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR5278-1G OR5278-100MG OR5278-250MG
77422-746EA 45 USD InStock
77422-746 77422-748 77422-750
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid
2-Phenylimidazo[1,2-a]pyridine-3-carboxylic acid


Formula: C₁₄H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00269226
CAS Number: 123533-41-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty