Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Cyano-4-methylaniline

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-Amino-2-methylbenzonitrile

OR5392-1G
77422-844EA 13 USD InStock
77422-844
3-Cyano-4-methylaniline
3-Cyano-4-methylaniline


Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 110 °C/22 mmHg
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00017608
CAS Number: 50670-64-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty