Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4,6-Dichloro-o-toluidine 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2,4-Dichloro-6-methylaniline

OR6295-1G
77423-730EA 13 USD InStock
77423-730
4,6-Dichloro-o-toluidine 97%
4,6-Dichloro-o-toluidine


Formula: C₇H₇Cl₂N
MW: 176.05 g/mol
Boiling Pt: 82…84 °C/0.1 mmHg
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD00044074
CAS Number: 30273-00-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty