Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dichloro-4-iodobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3,4-Dichloroiodobenzene

OR6486-100G OR6486-500G OR6486-25G
77423-808EA 60 USD InStock
77423-808 77423-806 77423-804
1,2-Dichloro-4-iodobenzene 99%
1,2-Dichloro-4-iodobenzene


Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
Boiling Pt: 116…118 °C/12 mmHg
Melting Pt: 27…29 °C
Storage Temperature: Ambient
MDL Number: MFCD00019014
CAS Number: 20555-91-3
EINECS: 243-874-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty