Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydrazinylpyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-hydrazinylpyridine2-Hydrazinopyridine2(1H)-Pyridinone, hydrazone

OR61068-5G OR61068-25G OR61068-100G
77423-476EA 28 USD InStock
77423-476 77423-474 77423-472
2-Hydrazinylpyridine 95%
2-Hydrazinylpyridine


Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 90…92 °C/0.75 mmHg
Melting Pt: 41…44 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006249
CAS Number: 4930-98-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty