Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-Methyl-2-amino-4-methylpentanoate hydrochloride

Supplier: APOLLO SCIENTIFIC
Synonyms: H-Leu-OMe.HClL-Leucine methyl ester hydrochlorideH-Leu-OMe·HCl

OR5853-25G OR5853-100G
77423-246EA 16 USD InStock
77423-246 77423-244
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride
(S)-Methyl-2-amino-4-methylpentanoate hydrochloride


Formula: C₇H₁₅NO₂·HCl
MW: 181.7 g/mol
Melting Pt: 145…155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012494
CAS Number: 7517-19-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty