Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-(trifluoromethoxy)benzoic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC1100BD-5G PC1100BD-25G PC1100BD-1G
77426-790EA 34 USD InStock
77426-790 77426-786 77426-788
4-Amino-3-(trifluoromethoxy)benzoic acid 97%
4-Amino-3-(trifluoromethoxy)benzoic acid


Formula: C₈H₆F₃NO₃
Melting Pt: 168…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00205143
CAS Number: 175278-22-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty