Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromotetrafluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Bromo-2,3,5,6-tetrafluoroaniline

PC1539-1G PC1539-5G PC1539-25G
77427-146EA 39 USD InStock
77427-146 77427-144 77427-142
4-Bromotetrafluoroaniline 98%
4-Bromotetrafluoroaniline


Formula: C₆H₂BrF₄N
MW: 243.9844 g/mol
Melting Pt: 59…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012144
CAS Number: 1998-66-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty