Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Bromo-2'-fluoroacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-(4-Bromo-2-fluorophenyl)ethanone

PC2391-25G PC2391-5G PC2391-1G
77427-646EA 28 USD InStock
77427-646 77427-644 77427-642
4'-Bromo-2'-fluoroacetophenone 98%
4'-Bromo-2'-fluoroacetophenone


Formula: C₈H₆BrFO
MW: 217.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411551
CAS Number: 625446-22-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty