Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-2-(trifluoromethoxy)toluene 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC2457-1G PC2457-250MG PC2457-5G
77427-658EA 503 USD InStock
77427-658 77427-656 77427-654
5-Bromo-2-(trifluoromethoxy)toluene 95%
5-Bromo-2-(trifluoromethoxy)toluene


Formula: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD04972695
CAS Number: 887268-26-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty