Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Difluorophenylhydrazine hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (3,5-Difluorophenyl)hydrazine hydrochloride

PC2721-25G PC2721-5G
77427-792EA 13 USD InStock
77427-792 77427-790
3,5-Difluorophenylhydrazine hydrochloride 97%
3,5-Difluorophenylhydrazine hydrochloride


Formula: C₆H₇Cl₃N₂
MW: 213.49 g/mol
Melting Pt: 261…266 °C
Storage Temperature: Ambient
MDL Number: MFCD03094171
CAS Number: 502496-27-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty