Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorocinnamic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-4-fluorophenyl acrylic acidtrans-4-Fluorocinnamic acid(2E)-3-(4-fluorophenyl)prop-2-enoic acidp-Fluorocinnamic acid

PC3732-500G PC3732-1KG PC3732-100G
77428-922EA 30 USD InStock
77428-922 77428-924 77428-926
4-Fluorocinnamic acid 99%
4-Fluorocinnamic acid


Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00004395
CAS Number: 459-32-5
EINECS: 207-288-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty