Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-3-nitrotoluene 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC4045-25G PC4045-100G
77429-312EA 34 USD InStock
77429-312 77429-314
4-Fluoro-3-nitrotoluene 99%
4-Fluoro-3-nitrotoluene


Formula: C₇H₆FNO₂
Boiling Pt: 241 °C
Melting Pt: 22…30 °C
Density: 1.262 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007060
CAS Number: 446-11-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty