Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorobenzyl alcohol 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (3-Fluorophenyl)methanol

PC3650-25G
77428-902EA 17 USD InStock
77428-902
3-Fluorobenzyl alcohol 98%
3-Fluorobenzyl alcohol


Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 201 °C
Density: 1.166 g/cm³ (25 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00004631
CAS Number: 456-47-3
EINECS: 207-265-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty