Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Heptafluorobutyro)phenone 97%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2,2,3,3,4,4,4-Heptafluorobutyrophenone

PC4483-25G PC4483-5G
77429-778EA 56 USD InStock
77429-778 77429-776
(Heptafluorobutyro)phenone 97%
(Heptafluorobutyro)phenone


Formula: C₁₀H₅F₇O
MW: 274.14
Boiling Pt: 170…172 °C
Density: 1.473 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077552
CAS Number: 559-91-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty