Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium 3,4-difluorophenylacetate

Supplier: APOLLO SCIENTIFIC
Warning

PC450193-1G PC450193-5G
77429-866EA 89 USD InStock
77429-866 77429-868
Sodium 3,4-difluorophenylacetate
Sodium 3,4-difluorophenylacetate


Formula: C₈H₅F₂O₂·Na
MW: 194.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28054313
CAS Number: 1980064-70-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty