Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-4-fluorobenzene -1,2-diamine 95%

Supplier: APOLLO SCIENTIFIC
Synonyms: 3-Chloro-4-fluorobenzene-1,2-diamine

PC501050-250MG PC501050-1G
77430-720EA 211 USD InStock
77430-720 77430-718
3-Chloro-4-fluorobenzene -1,2-diamine 95%
3-Chloro-4-fluorobenzene -1,2-diamine


Formula: C₆H₆ClFN₂
MW: 160.577 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18804697
CAS Number: 132915-81-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty