Would you like to log in again?
MegaMag Genie™ Magnetic Stirrers, 230 V
Supplier: Electron Microscopy Sciences
72404-AU 72404-BR 72404-EU 72404-20
102098-036EA 942.77 USD InStock
102098-036 102098-034 102098-032 102098-030
MegaMag Genie™ Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
The revolutionary single-position MagStir Genie provides low to high speed stirring and editable memory profiles
  • 230 V model
MagStir Genie may be used with any regular flask/beaker or spinner flask and a maximum sample weight of 5.0 kg. Speed range of 10 to 1800 rpm including reverse and interval stirring for applications ranging from gentle stirring for cell culture to aggressive mixing for viscous polymers. Three power levels are available for various sample viscosities. Low-profile magnetic stirrer uses microprocessor control for precise and reproducible operation without heat build-up from internal friction.
Manual mode provides stir, stir reverse, and time functions. Profiles mode allows users to quickly program speed, ramp rate, stir direction, time (including pause), and power level in up to 9 distinct steps. Up to 4 stirring profiles can be saved.
Automatic mode recalls and executes one of up to 4 user-defined profiles.
Microprocessor controls offer precision and reproducibility without internal heat build-up.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (102098-028) is also available, please see separate product page.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty