Would you like to log in again?
Metzenbaum Scissors,Tungsten Carbide, Integra™ Miltex®
Supplier: Integra LifeSciences
5-184TC 5-179TC
95039-252EA 449.32 USD InStock
95039-252 21909-160
Metzenbaum Scissors,Tungsten Carbide, Integra™ Miltex®
Scissors
Standard pattern
Stainless steel with tungsten carbide inserts in blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty