Would you like to log in again?
Eye Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
66-3240
82030-304EA 220.11 USD InStock
82030-304
Eye Tissue Forceps, OR Grade, Sklar
Forceps
Iris Tissue Forceps are used for grasping and handling tissue in ophthalmic procedures.
The narrow tips allow this instrument to be beneficial for use in other microsurgical procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty