Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

QuadMag Genie® Four-Position Magnetic Stirrers, 230 V

Supplier: Scientific Industries
SI-5246
89202-328EA 2073.55 USD InStock
89202-328
QuadMag Genie® Four-Position Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
These four-position magnetic stirrers are available in an analog model or digital model.

  • 230 V model

Choose the analog model, which is optimized for high speed, or choose the digital model for better speed control; this model offers digital display, control of speed, and is time-speed adjustable in 10 rpm increments. Digital models also incorporate a timer from 1 to 99 minutes and a ramping feature that slowly increases speed to avoid splashing and improve spin bar control. Can be used in cold rooms and incubators.

Powerful, general purpose four-position magnetic stirrers feature "rare earth" magnets that stay coupled under the most demanding strirring conditions and offer exceptional magnetic force capable of stirring up to 2 L for each position

Certifications: CE marked.

Ordering information: 230V models are supplied without a plug.

Caution: 220 V model. Corresponding model with voltage suitable for use in US (89202-326) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty