Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eurostar 40 Digital Electronic Overhead Stirrer, 230 V

Supplier: IKA
4444000
89412-488EA 2793.06 USD InStock
89412-488
Eurostar 40 Digital Electronic Overhead Stirrer, 230 V
Stirrers Overhead Stirrers
Universal laboratory stirrer designed for simple stirring tasks for quantities up to 25L (H₂O)

  • 230 V model

The Eurostar 40 Digital Electronic Overhead Stirrer features digital speed display, infinitely adjustable speed, push-through agitator shafts, overload protection, short-term overload operation, slim casing, quiet operation, and error code display.

The unit automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/30 to 2000 rpm. Safety circuits installed ensure automatic cut-off in anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed, even with changes in viscosities of the sample.

Certifications: CE marked.

Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration). Unit includes chuck, chuck key, and power cord. Stirring elements, boss head clamps, stands, and other accessories are sold separately.

Caution: 230 V model. Corresponding model with voltage suitable for use in US (89412-486) is also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty