Would you like to log in again?
C-MAG HS 4 Digital IKAMAG® Hotplate Stirrer, 230 V
Supplier: IKA
IKAMAG®
4240200
89495-306EA 1096.62 USD InStock
89495-306
C-MAG HS 4 Digital IKAMAG® Hotplate Stirrer, 230 V
Stirrers Magnetic Hotplate Stirrers
New magnetic stirrer features heating capabilities and a ceramic heating plate that offers excellent chemical resistance.
  • 230 V model
Units are equipped with a powerful motor for stirring quantities up to 5 L (H₂O). The LCD display provides simultaneous digital display of target and actual temperatures. Direct connection for a PT 1000 temperature sensor enables precise temperature control (included in delivery). Control accuracy in medium ±0.5K (in combination with PT 1000). Display of actual temperature value in medium with 0.1K resolution when using PT 1000 temperature sensor. Units offer three operating modes to choose from (Standard, Safe, Adjustment Protection). Standard safety features include a fixed safety circuit of 550 °C, Hot Top indicator for hot surface warning to prevent burns, a digital error code display, and an elevated control panel for protection against leaks.
Units meet IP 21 protection class according to DIN EN 60529.
Certifications: CE marked.
Ordering information: Supplied with three-year manufacturer's warranty (2+1 with product registration at www.ika.com/register). PT 1000 temperature sensor is included in delivery. For additional laboratory accessory options, see the IKA® Lablift Manual (see 89495-302 series). Other accessories are sold separately.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89495-304) is also available, please see separate product page.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty