Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dibromobutane

Supplier: APOLLO SCIENTIFIC

OR323244-100G OR323244-500G OR323244-1KG
77409-196EA 16 USD InStock
77409-196 77409-198 77409-200
1,4-Dibromobutane
1,4-Dibromobutane


Formula: BrH₂C(CH₂)₂CH₂Br
MW: 215.92 g/mol
Boiling Pt: 63…65 °C
Melting Pt: –20 °C
Density: 1.808 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000261
CAS Number: 110-52-1
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty