Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromoacetaldehyde diethyl acetal 97%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2-Bromo-1,1-diethoxyethane

OR28607-100G
77408-118EA 23 USD InStock
77408-118
Bromoacetaldehyde diethyl acetal 97%
Bromoacetaldehyde diethyl acetal


Formula: BrH₂CCH(OCH₂CH₃)₂
MW: 197.07 g/mol
Boiling Pt: 66…67 °C/18 mmHg
Density: 1.31 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000214
CAS Number: 2032-35-1
EINECS: 217-989-1
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty