Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-1-propanol

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Chloropropan-1-ol1-Chloro-3-hydroxypropane

OR0977-100G OR0977-25G OR0977-500G
77402-366EA 206 USD InStock
77402-366 77402-364 77402-362
3-Chloro-1-propanol
3-Chloro-1-propanol


Formula: C₃H₇ClO
MW: 94.54 g/mol
Boiling Pt: 160…162 °C
Melting Pt: –20 °C
Density: 1.131 g/cm³ (25 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00002943
CAS Number: 627-30-5
EINECS: 210-992-9
UN: 2849
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty