Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Iodoperfluoropropane 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-IodoheptafluoropropaneHeptafluoro-2-iodopropane1,1,1,2,3,3,3-Heptafluoro-2-iodopropaneHeptafluoroisopropyl iodide

PC4920-10G PC4920-25G PC4920-100G
77430-402EA 30 USD InStock
77430-402 77430-394 77430-396
2-Iodoperfluoropropane 98%
2-Iodoperfluoropropane


Formula: C₃F₇I
MW: 295.93 g/mol
Boiling Pt: 38 °C
Melting Pt: –56 °C
Density: 2.1 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00042089
CAS Number: 677-69-0
EINECS: 211-643-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty