Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trifluoroiodomethane 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC7680-25G PC7680-250G
77418-116EA 375 USD InStock
77418-116 77418-118
Trifluoroiodomethane 99%
Trifluoroiodomethane
Refrigerate before opening.


Formula: CF₃I
MW: 195.91 g/mol
Boiling Pt: –22.5 °C
Density: 2.36 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001060
CAS Number: 2314-97-8
EINECS: 219-014-5
UN: 1956
ADR: 2.2,
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty