Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diiodomethane 98+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Methylene iodide

OR17001-100G
77405-896EA 127 USD InStock
77405-896
Diiodomethane 98+%
Diiodomethane


Formula: CH₂I₂
MW: 267.84 g/mol
Boiling Pt: 180…181 °C
Melting Pt: 5...7 °C
Density: 3.322 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001079
CAS Number: 75-11-6
EINECS: 200-841-5
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty