Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3-chlorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 3-Bromochlorobenzene

OR1470-100G OR1470-25G
77405-074EA 31 USD InStock
77405-074 77405-076
1-Bromo-3-chlorobenzene 98%
1-Bromo-3-chlorobenzene


Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 195…197 °C
Melting Pt: –22 °C
Density: 1.63 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00000568
CAS Number: 108-37-2
EINECS: 203-575-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty