Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vanillic acid

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 4-Hydroxy-3-methoxybenzoic acid

OR1181-100G OR1181-5G OR1181-25G
77404-038EA 20 USD InStock
77404-038 77404-040 77404-036
Vanillic acid
Vanillic acid


Formula: C₈H₈O₄
MW: 168.15 g/mol
Melting Pt: 210…213 °C
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty