Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2-fluoroaniline 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC1819W-25G PC1819W-500G PC1819W-100G
77413-320EA 17 USD InStock
77413-320 77413-322 77413-324
3-Chloro-2-fluoroaniline 97%
3-Chloro-2-fluoroaniline


Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 214 °C
Density: 1.324 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00069415
CAS Number: 2106-04-9
EINECS: 218-283-6
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty