Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Chloro-2-nitrobenzene 99%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1-Chloro-2-nitrobenzene2-Chloronitrobenzene

OR5399-250G OR5399-5KG OR5399-1KG
77410-354EA 313 USD InStock
77410-354 77410-352 77410-350
1-Chloro-2-nitrobenzene 99%
1-Chloro-2-nitrobenzene


Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 245…246 °C
Melting Pt: 31…34 °C
Density: 1.348 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007061
CAS Number: 88-73-3
EINECS: 201-854-9
UN: 1578
ADR: 6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty