Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perfluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: Octafluorotoluene1,2,3,4,5-Pentafluoro-6-(trifluoromethyl)benzeneα,α,α,2,3,4,5,6-OctafluorotoluenePentafluoro(trifluoromethyl)benzene

PC5354-100G PC5354-25G
77416-712EA 168 USD InStock
77416-712 77416-714
Perfluorotoluene 98%
Perfluorotoluene
In base, hydroxyl groups react at the 4-position to yield heptafluoro-p-tolyl ethers, which can be used to protect estradiols.


Formula: C₇F₈
MW: 236.06 g/mol
Boiling Pt: 103…105 °C
Melting Pt: –65.6 °C
Density: 1.666 g/cm³ (25 °C)
Flash Pt: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00000375
CAS Number: 434-64-0
EINECS: 207-104-7
UN: 1992
ADR: 3+6.1,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty