Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Chloro-m-xylene 94%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1-(Chloromethyl)-3-methylbenzene3-Methylbenzyl chloride

OR2562-100G OR2562-250G
77407-622EA 23 USD InStock
77407-622 77407-624
α-Chloro-m-xylene 94%
α-Chloro-m-xylene


Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 195…196 °C
Density: 1.064 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000909
CAS Number: 620-19-9
EINECS: 210-628-9
UN: 3265
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty