Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromocyclopentane

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Cyclopentyl bromide

OR10098-25G OR10098-250G OR10098-100G
77402-470EA 44 USD InStock
77402-470 77402-474 77402-472
Bromocyclopentane
Bromocyclopentane


Formula: C₅H₉Br
MW: 149.03 g/mol
Boiling Pt: 137…139 °C
Density: 1.39 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00001359
CAS Number: 137-43-9
EINECS: 205-294-6
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty