Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hexafluorophosphate, anhydrous 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC6510-100G
77417-416EA 121 USD InStock
77417-416
Sodium hexafluorophosphate, anhydrous 98%
Sodium hexafluorophosphate


Formula: F₆NaP
MW: 167.95 g/mol
Melting Pt: >200 °C
Density: 2.369 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011122
CAS Number: 21324-39-0
EINECS: 244-333-1
UN: 3260
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty