Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-fluoro-5-nitrobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Bromo-1-fluoro-4-nitrobenzene3-Bromo-4-fluoronitrobenzene

PC9956-25G
77434-660EA 16 USD InStock
77434-660
1-Bromo-2-fluoro-5-nitrobenzene 98%
1-Bromo-2-fluoro-5-nitrobenzene


Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 55…58 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00055433
CAS Number: 701-45-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty