Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromoresorcinol 95%

Supplier: APOLLO SCIENTIFIC
Synonyms: 4-Bromobenzene-1,3-diol

OR480711-5G OR480711-25G OR480711-10G
77422-162EA 83 USD InStock
77422-162 77422-164 77422-166
4-Bromoresorcinol 95%
4-Bromoresorcinol


Formula: C₆H₅BrO₂
MW: 189.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002272
CAS Number: 6626-15-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty